
Methanesulphonic anhydride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7143-01-3
:
Formula:C₂H₆O₅S₂
MDL Number:MFCD00007556
:174.2 g/mol
Boiling Pt:125 °C/4mmHg
Melting Pt:64…67 °C
Storage Temperature:Ambient
EINECS:230-442-1
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together