4-(Cyanomethyl)benzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:50685-26-2
:
Formula:C₉H₇NO₂
MDL Number:MFCD00020382
Melting Pt:196 °C
:161.16 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together