Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate
Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate
Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate
Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate
APOSOR15094-1G
:
Apollo Scientific
:
Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate
APOSOR15094-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
218.21 g/mol
:
189…193 °C
:
GHS07
:
Ambient
:
29113-33-5
:
C₁₁H₁₀N₂O₃
:
MFCD00051583