Order Entry
France
ContactUsLinkComponent
2,5-Dibromothiophene > 98%
Toxic
Health
2,5-Dibromothiophene > 98%
Catalog # APOSOR14924-100G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
Health
2,5-Dibromothiophene > 98%
Catalog # APOSOR14924-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :241.93 g/mol
Boiling Pt:210…212 °C
Melting Pt:–6 °C
Density:2.147 g/cm³ (25 °C)
Flash Pt:99 °C
Storage Temperature:Refrigerator
EINECS:221-547-3
UN:2810
ADR:6.1,II
 :GHS06
 :GHS08
CAS Number:3141-27-3
Formula:C₄H₂Br₂S
MDL Number:MFCD00005420