4-Acetyltetrahydro-1(2H)-pyrazinecarboximidamidehydroiodide
Catalog # APOSOR1484-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:298.13 g/mol
Melting Pt:229…230 °C
:GHS07
Storage Temperature:Ambient
CAS Number:849776-26-7
Formula:C₇H₁₅IN₄O
MDL Number:MFCD06797494