Fmoc-L-4-pyridylalanine 98%
Catalog # APOSOR14664-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: N-Fmoc-3-(4-pyridyl)-L-alanine, Fmoc-3-(4-pyridyl)-Ala-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(4-pyridyl)-L-alanine, Fmoc-4-Pal-OH
:
:388.43 g/mol
Storage Temperature:Refrigerator
CAS Number:169555-95-7
Formula:C₂₃H₂₀N₂O₄
MDL Number:MFCD00672566