Fmoc-2-methyl-L-phenylalanine 95+%
Catalog # APOSOR14566-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:401.47 g/mol
Storage Temperature:Ambient
CAS Number:211637-75-1
Formula:C₂₅H₂₃NO₄
MDL Number:MFCD00671391