10-Bromodecanoic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:50530-12-6
:
Formula:C₁₀H₁₉BrO₂
MDL Number:MFCD00014388
:251.16 g/mol
Boiling Pt:163…165 °C/2mmHg
Melting Pt:38…42 °C
Flash Pt:>231 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together