3-Cyano-5-methylpyridine
Catalog # APOSOR13616-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:118.14 g/mol
Melting Pt:83…87 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
CAS Number:42885-14-3
Formula:C₇H₆N₂
MDL Number:MFCD00673151