
Benzotriazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:95-14-7
:
Formula:C₆H₅N₃
MDL Number:MFCD00005699
:119.13 g/mol
Boiling Pt:204 °C/15mmHg
Melting Pt:96…99 °C
Density:1.36 g/cm³ (25 °C)
Flash Pt:170 °C
EINECS:202-394-1
:GHS07
Storage Temperature:Ambient
Frequently Bought Together