
5-Methylthiophene-2-carbonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:31555-59-6
:
Formula:C₆H₅ClOS
MDL Number:MFCD00130092
:160.615 g/mol
Boiling Pt:120 °C/8mmHg
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together