Order Entry
France
Orders LinkContactUsLinkComponent
 

3-Hydroxyisonicotinic acid

Irritant
Supplier:   Apollo Scientific
3-Hydroxyisonicotinic acid
Supplier:  Apollo Scientific
Certificates
3-Hydroxyisonicotinic acid

3 Options Available Below

Product Details & Documents
Synonyms:  3-Hydroxypyridine-4-carboxylic acid
  • CAS Number:10128-71-9
  • Formula:C₆H₅NO₃
  • MDL Number:MFCD00234165
  • Molecular Weight:139.11 g/mol
  • Melting Pt:315…319 °C (dec.)
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 37,70 €
    37,70 €
    1 * 5 g
       37,70 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 118,00 €
    118,00 €
    1 * 25 g
       118,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.