
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:529-34-0
:
Formula:C₁₀H₁₀O
MDL Number:MFCD00001688
:146.19 g/mol
Boiling Pt:127 °C
Melting Pt:2 °C
Density:1.096 g/cm³ (25 °C)
Flash Pt:129 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together