Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dimethyl-2-(4-nitrophenyl)malonate
Dimethyl-2-(4-nitrophenyl)malonate
Dimethyl-2-(4-nitrophenyl)malonate
APOSOR110186-1G
:
Apollo Scientific
:
Dimethyl-2-(4-nitrophenyl)malonate
APOSOR110186-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
253.21 g/mol
:
Ambient
:
4033-88-9
:
C₁₁H₁₁NO₆
:
MFCD03425806