Methyl-6-(1H-pyrrol-1-yl)nicotinate
Catalog # APOSOR110054-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:202.21 g/mol
Storage Temperature:Ambient
CAS Number:852180-80-4
Formula:C₁₁H₁₀N₂O₂
MDL Number:MFCD04973730