Order Entry
France
Orders LinkContactUsLinkComponent
3,4-Dichlorobenzaldehyde 97%

3,4-Dichlorobenzaldehyde 97%

Corrosive
Supplier:   Apollo Scientific
3,4-Dichlorobenzaldehyde 97%
Supplier:  Apollo Scientific
Certificates
3,4-Dichlorobenzaldehyde 97%

2 Options Available Below

Product Details & Documents
  • CAS Number:6287-38-3
  • Formula:C₇H₄Cl₂O
  • MDL Number:MFCD00003351
  • Molecular Weight:175.01 g/mol
  • Boiling Pt:247…248 °C
  • Melting Pt:39…42 °C
  • Flash Pt:>110 °C
  • Storage Temperature:Refrigerator
  • EINECS:228-520-5
  • UN:1759
  • ADR:8,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 30,40 €
    30,40 €
    1 * 25 g
       30,40 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 88,30 €
    88,30 €
    1 * 100 g
       88,30 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.