
2,4-Dibromothiophene, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3140-92-9
:
Formula:C₄H₂Br₂S
MDL Number:MFCD00043889
:241.93 g/mol
Boiling Pt:215 °C
Density:2.157 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Refrigerator
UN:2810
ADR:6.1,III
:GHS07
Frequently Bought Together