

(Methoxymethyl)triphenylphosphonium chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4009-98-7
:
Formula:C₂₀H₂₀ClOP
MDL Number:MFCD00011800
:342.8 g/mol
Melting Pt:185…195 °C (dec.)
Storage Temperature:Ambient
EINECS:223-664-5
UN:3077
ADR:9+EHS,III
:GHS07
:GHS09
Frequently Bought Together