2-Methylcyclohexanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:583-59-5
:
Formula:C₇H₁₄O
MDL Number:MFCD00001443
:114.19 g/mol
Boiling Pt:163…167 °C
Melting Pt:–21 °C
Density:0.929 g/cm³ (25 °C)
Flash Pt:58 °C
Storage Temperature:Ambient
EINECS:209-512-0
UN:2617
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together