

Sodium iodide, anhydrous 99+%
:7681-82-5
:
:NaI
:MFCD00003532
:149.89 g/mol
:1304 °C
:661 °C
:3.67 g/cm³ (25 °C)
:Ambient
:231-679-3
:3077
:9+EHS,III
:GHS07
:GHS09
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
1,3,4,6-Tetrachloro-3-6-diphenylglycouril
: Apollo Scientific
Sodium chlorite tech. ∼80%, Technical Grade
: Thermo Fisher Scientific