Order Entry
France
ContactUsLinkComponent
DIM-C-pPhCO2Me 99%
DIM-C-pPhCO2Me 99%
Catalog # APOSBISN0093-1MG
Supplier:  Apollo Scientific
CAS Number:  
DIM-C-pPhCO2Me 99%
Catalog # APOSBISN0093-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :380.45 g/mol
Storage Temperature:Refrigerator
CAS Number:151358-48-4
Formula:C₂₅H₂₀N₂O₂
MDL Number:MFCD31697720