

Levofloxacin
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:100986-85-4
:
Formula:C₁₈H₂₀FN₃O₄
MDL Number:MFCD00865049
Boiling Pt:572 °C
Melting Pt:218…227 °C
Flash Pt:>110 °C
:361.37 g/mol
:GHS07
:GHS08
Storage Temperature:Ambient
Frequently Bought Together