

5-Aza-2'-deoxycytidine free base
:2353-33-5
:
:C₈H₁₂N₄O₄
:MFCD00043011
:193…203 °C
:228.21 g/mol
:GHS07
:GHS08
:Freezer
Frequently Bought Together
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
: VWR Chemicals
Methanol ≥99.90%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
: Honeywell Chemicals
Acetone ≥99.0% Ph. Eur.
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
n-Heptane 99+% for HPLC
: Thermo Fisher Scientific