Order Entry
France
ContactUsLinkComponent
Glycerine, anhydrous, Cell Culture Grade
Irritant
Glycerine, anhydrous, Cell Culture Grade
Catalog # APOSBIA3092-500ML
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Glycerine, anhydrous, Cell Culture Grade
Catalog # APOSBIA3092-500ML
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :92.09 g/mol
Boiling Pt:182 °C/20mmHg
Melting Pt:20 °C
Density:1.25 g/cm³ (25 °C)
Flash Pt:160 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:56-81-5
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722