N-Acetyl-DL-tryptophan
Catalog # APOSBIA1791-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:246.27 g/mol
Storage Temperature:Freezer
CAS Number:87-32-1
Formula:C₁₃H₁₄N₂O₃
MDL Number:MFCD00005644