Order Entry
France
ContactUsLinkComponent
Mouse recombinant PD-L1 (from HEK293)
Mouse recombinant PD-L1 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Mouse recombinant PD-L1 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 0,1 EU per μg by the LAL method
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation; Measured by its binding ability in a functional ELISA. immobilised Mouse B7-1, Fc Tag (Cat. No. ACRBCD0-M5259) at 5 μg/ml (100 μl/well) can bind Mouse PD-L1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. ACRBPD1-M52A2) with a linear range of 0,16 - 2,5 μg/ml (QC tested); Measured by its binding ability in a functional ELISA. immobilised Mouse PD-L1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. ACRBPD1-M52A2) at 5 μg/ml (100 μl/well) can bind Biotinylated Mouse PD-1, Fc,Avitag (Cat. No. ACRBPD1-M82F4) with a linear range of 0,313 - 5 μg/ml (Routinely tested)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    B7-H1|PD-L1|CD274|PDCD1L1|PDCD1LG1
  • Protein/peptide name:
    PD-L1
  • Purity:
    >95%
  • Molecular weight:
    51,8 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

Specifications

Frequently Bought Together