Rabbit recombinant PD-L1 (from HEK293 cells)
Catalog # ACRBPDL-R52H6100UG
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Pk:100 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Rabbit
- Tag sequence:His Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Gene ID:G1SUI3-1
- Protein synonyms:B7-H1|PD-L1|CD274|PDCD1L1|PDCD1LG1
- Protein/peptide name:PD-L1
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:27,1 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
- Shipping temperature:RT
Specifications
About this item
Rabbit PD-L1 / B7-H1 protein, His Tag.