Mouse recombinant FGF R4 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyopilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Mouse FGF R4, His Tag at 2 μg/ml (100 µl/well) can bind Human FGF-9, Fc Tag with a linear range of 0,156-5 μg/ml (QC tested).
- Gene ID:Q03142-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:TKF|FGFR4|JTK2|CD334|MGC20292
- Protein/peptide name:FGF R4
- Purity:>90%
- Molecular weight:40,7 kDa
- Endotoxin level:Low
- Formulation:Lyopilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together