Human recombinant TNFSF11 (from HEK293)
Catalog # ACRBRAL-H52A6-1MG
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Pk:1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Seq:02
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyopilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Captured Human RANK, Mouse IgG2a Fc Tag, low endotoxin on CM5 chip via Anti-Mouse antibodies surface can bind Human TNFSF11, Fc Tag with an affinity constant of 0,574 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; Loaded Human RANK, Mouse IgG2a Fc Tag, low endotoxin on AMC Biosensor, can bind Human TNFSF11, Fc Tag with an affinity constant of 0,205 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:OPGL|ODF|TRANCE|CD254|RANKL
- Protein/peptide name:TNFSF11
- Purity:>90%
- Molecular weight:46,9 kDa
- Endotoxin level:Low
- Formulation:Lyopilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT