Order Entry
France
ContactUsLinkComponent
Human recombinant integrin alpha 8 beta 1 (from HEK293)
Human recombinant integrin alpha 8 beta 1 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant integrin alpha 8 beta 1 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised fibronectin at 5 μg/ml (100 μl/well) can bind biotinylated human ITGA8 and ITGB1 heterodimer protein, His,Avitag and tag free (Cat. No. ACBRIT1-H82Wb) with a linear range of 0,156 - 5 μg/ml (QC tested).
  • Gene ID:
    P53708-1 (ITGA8) and P05556-1 (ITGB1)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    Integrin alpha 8 beta 1|ITGA8 and ITGB1
  • Protein/peptide name:
    Integrin alpha 8 beta 1
  • Purity:
    >90%
  • Molecular weight:
    116,4 kDa (ITGA8) and 83,7 kDa (ITGB1)
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together