Human recombinant CD8 alpha & beta (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human CD8 alpha&beta (CD8A&CD8B) Heterodimer Protein, His Tag&Tag Free (MALS verified), ACROBiosystems.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag&Tag Free (MALS verified)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Gene ID:P01732-1 (CD8A) & P10966-1 (CD8B)
- Protein synonyms:CD8A & CD8B|CD8 alpha & beta
- Protein/peptide name:CD8 alpha & beta
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:19,5 kDa (CD8A) & 22,1 kDa (CD8B)
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together