Order Entry
France
ContactUsLinkComponent
 
Rat recombinant CTLA-4 (from HEK293)
 
Rat recombinant CTLA-4 (from HEK293)

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rat
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with glycine, arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Gene ID:
    Q62859
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CTLA4|CD152
  • Protein/peptide name:
    CTLA-4
  • Purity:
    >95%
  • Molecular weight:
    40,2 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with glycine, arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

 

Frequently Bought Together