Mouse recombinant OX40 ligand (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:−20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised mouse OX40 ligand, His Tag (Cat. No. ACRBTN4-M5241) at 0,5 μg/ml (100 μl/well) can bind mouse OX40, Fc Tag (Cat. No. ACRBOX0-M5259) with a linear range of 10 - 16 ng/ml (QC tested).
- Gene ID:P43488-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:TNFSF4|CD134L|TXGP1|OX40L|Glycoprotein Gp34|CD134 ligand|CD252
- Protein/peptide name:OX40 Ligand
- Purity:>90%
- Molecular weight:18,3 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together