Order Entry
France
ContactUsLinkComponent
 
Mouse recombinant serum albumin (from HEK293 cells)
 
Mouse recombinant serum albumin (from HEK293 cells)

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Mouse Serum Albumin, His Tag (Cat. No. MSA-M52H8) Immobilised on CM5 Chip can bind Mouse FCGRT&B2M Heterodimer Protein, His Tag (Cat. No. FCM-M52W8) with an affinity constant of 0,165 μM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded Biotinylated Mouse FCGRT&B2M Heterodimer Protein, His,Avitag (BLI verified)(Cat. No. FCM-M82W5) on SA Biosensor, can bind Mouse Serum Albumin, His Tag(Cat. No. MSA-M52H8) with an affinity constant of 0,495 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    P07724-1
  • Protein synonyms:
    Serum albumin|Alb|ALB
  • Protein/peptide name:
    Serum Albumin
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    67,8 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Shipping temperature:
    RT

 

Frequently Bought Together