Human recombinant IL-13 R alpha 2 (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag (MALS verified)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Gene ID:Q14627
- Protein synonyms:IL13RA2|CD213A2|IL-13R|CT19|IL13BP
- Protein/peptide name:IL-13 R alpha 2
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:63,5 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together