Order Entry
France
ContactUsLinkComponent
Human recombinant Fc gamma RIIA/ CD32a (from HEK293 cells)
Human recombinant Fc gamma RIIA/ CD32a (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant Fc gamma RIIA/ CD32a (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag (SPR & BLI verified)
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Immobilised Human CD32a (H167), His Tag (SPR & BLI verified) (Cat. No. CD1-H5223) on CM5 Chip via anti-His antibody, can bind  Rituximab with an affinity constant of 1,45 μM as determined in a SPR assay (Biacore T200) (QC tested). Loaded Human CD32a (H167), His Tag (SPR & BLI verified) (Cat. No. CD1-H5223) on HIS1K Biosensor, can bind Rituximab with an affinity constant of 1,30 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    P12318-1
  • Protein synonyms:
    CD32|CD32a|IGFR2|FCGR2A1|FCG2 |FCGR2A
  • Protein/peptide name:
    Fc gamma RIIA/ CD32a
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    21,5 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together