Order Entry
France
ContactUsLinkComponent
Human recombinant IL-1 Rrp2 (from HEK293)
Human recombinant IL-1 Rrp2 (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human recombinant IL-1 Rrp2 (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Recombinant Human IL-36Ra / IL-1F5 at 2 μg/ml (100 μl/well) can bind Human IL-1 Rrp2, Fc Tag (Cat. No. ACRBIL2-H5254) with a linear range of 0,156 - 5 μg/ml (QC tested).
  • Gene ID:
    Q9HB29-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    IL1RL2|IL-1RL2|IL-36R|IL-1Rrp2|IL1Rrp2|IL-1 R6|IL1RRP2|IL-1 Rrp2
  • Protein/peptide name:
    IL-1 Rrp2
  • Purity:
    >95%
  • Molecular weight:
    62,4 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together