Order Entry
France
ContactUsLinkComponent
Human recombinant Glypican 3 (from HEK293), Acro Biosystems
Human recombinant Glypican 3 (from HEK293), Acro Biosystems
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant Glypican 3 (from HEK293), Acro Biosystems
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 0,1 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human FGF basic, Tag Free (Cat. No. ACRBBFF-H4117) at 0.5 μg/ml (100 μl/well) can bind Human Glypican 3, His Tag (Cat. No. ACRBGP3-H52H4) with a linear range of 8 - 31 ng/ml (QC tested). Measured by its binding ability in a functional ELISA. Immobilised Human Glypican 3, His Tag (Cat. No. ACRBGP3-H52H4) at 0,5 μg/ml (100 μl/well) can bind Anti-Human GPC3 MAb with a linear range of 0,1 - 2 ng/ml (Routinely tested). Anti-human GPC3 MAb (chimeric mouse-human IgG1) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind Human Glypican 3, His Tag (Cat. No. ACRBGP3-H52H4) with an affinity constant of 1,33 nM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded Anti-Human GPC3 MAb (chimeric mouse-human IgG1) on AHC Biosensor, can bind Human Glypican 3, His Tag (Cat. No. ACRBGP3-H52H4) with an affinity constant of 1,48 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    P51654-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    GPC3|SGBS|SGB|Glypican-3|SDYS |MXR7|SGBS1|OCI5|GTR2-2|DGSX
  • Protein/peptide name:
    Glypican 3
  • Purity:
    >90%
  • Molecular weight:
    38,1 kDa and 24,6 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together