Human recombinant nectin-2 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,1 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before Lyophilisation.; Loaded Human Nectin-2, Mouse IgG2a Fc Tag (Cat. No. CD2-H5257) on Protein A Biosensor, can bind Human PVRIG, His Tag (Cat. No. PVG-H52H4) with an affinity constant of 1.33 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q92692-2
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:Nectin-2|PVRL2|CD112
- Protein/peptide name:Nectin-2
- Purity:>95%
- Molecular weight:62,4 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before Lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together