Human recombinant Mesothelin (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4, Normally trehalose is added as protectant before Lyophilisation,; 293 cells were transfected with anti-MSLN-scFv and RFP tag, 2e5 of the cells were first stained with B, Human Mesothelin (296-580), Fc Tag (Cat, No, MSN-H5253, 1 µg/ml) and C, Fc Tag Fusion Protein Control, followed by FITC anti-human IgG Fc, A, Non-transfected 293 cells and C, Fc Tag Fusion Protein Control were used as negative control, RFP was used to evaluate CAR (anti-MSLN-scFv) expression and FITC was used to evaluate the binding activity of Human Mesothelin (296-580), Fc Tag (Cat, No, MSN-H5253).
- Gene ID:Q13421-3 (R309P)
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:MPF|MSLN|Mesothelin
- Protein/peptide name:Mesothelin
- Purity:>95%
- Molecular weight:58,6 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4, Normally trehalose is added as protectant before Lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together