Order Entry
France
ContactUsLinkComponent
(2-Fluoro-benzoylamino)-acetic acid 98%
(2-Fluoro-benzoylamino)-acetic acid 98%
Catalog # APOSPC909033-1G
Supplier:  Apollo Scientific
CAS Number:  
(2-Fluoro-benzoylamino)-acetic acid 98%
Catalog # APOSPC909033-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :197.163 g/mol
Storage Temperature:Ambient
CAS Number:363-34-8
Formula:C₉H₈FNO₃
MDL Number:MFCD00460020