
Pentafluorophenyl nicotinate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:848347-44-4
:
Formula:C₁₂H₄F₅NO₂
MDL Number:MFCD09064938
:289.158 g/mol
Melting Pt:65.5…67 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together