4-Amino-2,5-difluorophenol
Catalog # APOSPC8310-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:145.107 g/mol
Melting Pt:125…130 °C
Flash Pt:>110 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS07
CAS Number:120103-19-7
Formula:C₆H₅F₂NO
MDL Number:MFCD03094492