Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-1-(2, 4-Difluorophenyl)ethylamine
(S)-1-(2, 4-Difluorophenyl)ethylamine 97%
(S)-1-(2, 4-Difluorophenyl)ethylamine 97%
APOSPC56179-250MG
:
Apollo Scientific
:
(S)-1-(2, 4-Difluorophenyl)ethylamine 97%
APOSPC56179-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
157.161 g/mol
:
Ambient
:
845252-02-0
:
C₈H₉F₂N
:
MFCD06761834