
Perfluorobutane-1-sulphonic anhydride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:36913-91-4
:
Formula:C₈F₁₈O₅S₂
MDL Number:MFCD03427256
:582.184 g/mol
Boiling Pt:84 °C
Density:1.898 g/cm³ (25 °C)
Storage Temperature:Refrigerator
EINECS:253-270-9
UN:3264
ADR:8,II
:GHS05
Frequently Bought Together