6-(Trifluoromethyl)pyridine-2-carboxamidine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:264884-49-3
:
Formula:C₇H₆F₃N₃·ClH
MDL Number:MFCD00662774
:225.599 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together