
(6-Bromo-3-fluoropyridin-2-yl)methanol
Catalog # APOSPC48635-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:206.012 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:918793-01-8
Formula:C₆H₅BrFNO
MDL Number:MFCD16607006