Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(R)-1-(3,5-Difluorophenyl)ethanol
(R)-1-(3,5-Difluorophenyl)ethanol
(R)-1-(3,5-Difluorophenyl)ethanol
APOSPC450096-1G
:
Apollo Scientific
:
(R)-1-(3,5-Difluorophenyl)ethanol
APOSPC450096-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
158.145 g/mol
:
GHS07
:
Ambient
:
433228-88-7
:
C₈H₈F₂O
:
MFCD18831076