Order Entry
France
ContactUsLinkComponent
[(1R)-1-(3,5-Difluorophenyl)ethyl][(1S)-1-phenylethyl]ammonium fumarate
[(1R)-1-(3,5-Difluorophenyl)ethyl][(1S)-1-phenylethyl]ammonium fumarate
Catalog # APOSPC450047-250MG
Supplier:  Apollo Scientific
CAS Number:  
[(1R)-1-(3,5-Difluorophenyl)ethyl][(1S)-1-phenylethyl]ammonium fumarate
Catalog # APOSPC450047-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :377.382 g/mol
Storage Temperature:Ambient
Formula:C₁₆H₁₇F₂N·C₄H₄O₄
MDL Number:MFCD25978113