Order Entry
France
ContactUsLinkComponent
(S)-3,3'-Bis(3,5-difluorophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
(S)-3,3'-Bis(3,5-difluorophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Catalog # APOSPC440007-250MG
Supplier:  Apollo Scientific
CAS Number:  
(S)-3,3'-Bis(3,5-difluorophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Catalog # APOSPC440007-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :572.442 g/mol
Storage Temperature:Ambient
Formula:C₃₂H₁₇F₄O₄P
MDL Number:MFCD28139328